Magicard™ Prima 8 Duo Laminator
Lieferant: Brady
321958
BRDY321958EA 2750 EUR
BRDY321958
Magicard™ Prima 8 Duo Laminator
Drucker Document Printers
The Magicard Prima 8 offers a reliable, robust solution for immediate card dispensing and lamination in a compact format.
  • Add this laminator to the Prima 8 Duo and print and laminate your cards on both sides in one go
  • Provide your card with extra protection by applying an overlaminate over the print
  • Visual scurity - holographic laminates and UV film options
Using retransfer printing, images are first printed onto a thin film and then heat-transferred onto the card surface. This creates a high-resolution, glossy image printed directly over the edge of the card.