Thermal transfer printer ribbon, R6606 series
Lieferant: Brady
622116
BRDY622116EA 78.59 EUR
BRDY622116
Thermal transfer printer ribbon, R6606 series
Farbbänder
Diesel-resistant thermal transfer ribbon on Brady PermaSleeve ™ and Heatex ™ heat shrink tubing. This print ribbon is highly resistant to chemicals and friction.
Verpackung: One print ribbon/box.