Ground socket
Lieferant: Brand
51467
BRND51467EA 26.4 EUR
BRND51467
Ground socket
Adapter Übergangsstücke mit Kegelschliffen
PTFE. With grip collar and sealing rings for excellent tightness.
  • Greaseless seal will not contaminate sample
  • Outstanding chemical resistance at operating temperatures from −200 to +260 °C
  • Sturdy design for permanent use
Low leakage rate: Q < 1 · 10-4 mbar · l · s-1.