SamplePro Needle Mounting Adapter
Lieferant: BRUKER DALTRONIK
H160098
BRUKH160098EA 187 EUR
BRUKH160098
SamplePro Needle Mounting Adapter
Injektionsnadeln
SPT NADELINSERT KPL is required to mount new needle version in the SamplePro Tube liquid Handler.
Suitable for needles H160071, H160072, H160073. One insert per SamplePro Tube required.