Sealing tape

Lieferant: BURKLE
0006-4514
BURK0006-4514EA 2.56 EUR
BURK0006-4514
Sealing tape
Klebebänder
This PTFE sealing tape is ideal for sealing threads.

  • Low abrasion
  • Excellent chemical resistance
  • Temperature resistant from −80 to +260 °C

Length: 12,0 m

Colour: White
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR