Calcein AM 4 mM in DMSO ≥90% (durch HPCE), klare Lösung

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Calcein-AM, Calcein O,O′-diacetate tetrakis(acetoxymethyl) ester, Calcein acetoxymethyl ester, Tetrakis(acetoxymethyl) 2,2',2'',2'''-(((3',6'-diacetoxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetate

C1359-100UL
C1359-100ULEA 428 EUR
C1359-100UL
Calcein AM 4 mM in DMSO ≥90% (durch HPCE), klare Lösung
Calcein AM
Formel: C₄₆H₄₆N₂O₂₃
Molecular Weight: 994,86 g/mol
MDL: MFCD05861516
CAS-Nummer: 148504-34-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ad2c549f73e983d4138d70c9503daa941504371&vid=6eb46d48d455869b8a0c4691139901db686061d3&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Colorless and White to Off White
Appearance (Form) Liquid
Wavelength
Lambda Max, before hydrolysis
≤300 nm
Purity (HPLC) ≥90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR