5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)
Lieferant: Cayman Chemical
|
Achtung
|
Synonyme:
Bis(4-nitrophenyl)disulfid-3,3'-dicarbonsäure, DTNB, 2,2'-Dinitro-5,5'-dithiodibenzoesäure, Ellman's Reagenz, 3,3'-Dithiobis(6-nitrobenzoesäure)
703012-1
CAYM703012-1EA
22
EUR
CAYM703012-1
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)
Formel:
C₁₄H₈N₂O₈S₂ Molecular Weight: 396,36 g/mol Schmelzpunkt: 240…244 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007140 CAS-Nummer: 69-78-3 EINECS: 200-714-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...