5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)

Lieferant: Cayman Chemical
Achtung

Synonyme: Bis(4-nitrophenyl)disulfid-3,3'-dicarbonsäure, DTNB, 2,2'-Dinitro-5,5'-dithiodibenzoesäure, Ellman's Reagenz, 3,3'-Dithiobis(6-nitrobenzoesäure)

765012-1
CAYM765012-1EA 14 EUR
CAYM765012-1
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)
Formel: C₁₄H₈N₂O₈S₂
Molecular Weight: 396,36 g/mol
Schmelzpunkt: 240…244 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007140
CAS-Nummer: 69-78-3
EINECS: 200-714-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR