4-Amino-3,5-dichlorbenzonitril
Lieferant: CHEMPUR
BTB03631.250
BTB03631.1
CHMPBTB03631.250EA
0
EUR
CHMPBTB03631.250
MAYBBTB03631.1
4-Amino-3,5-dichlorbenzonitril
4-Amino-3,5-dichlorbenzonitril
Formel:
C₇H₄Cl₂N₂ Molecular Weight: 187,03 g/mol Siedepunkt: 323 °C (1013 hPa) Schmelzpunkt: 120 °C |
MDL:
MFCD00017341 CAS-Nummer: 78473-00-4 EINECS: 278-914-6 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...