1,2-Benzoldisulfonylchlorid

Lieferant: CHEMPUR

Synonyme: Benzol-1,2-disulfonylchlorid, Benzol-1,2-disulfonsäurechlorid, 1,2-Benzoldisulfonsäurechlorid

BTB09872.250 BTB09872.1
CHMPBTB09872.250EA 0 EUR
CHMPBTB09872.250 MAYBBTB09872.1
1,2-Benzoldisulfonylchlorid
1,2-Benzoldisulfonylchlorid
Formel: C₆H₄Cl₂O₄S₂
Molecular Weight: 275,13 g/mol
MDL: MFCD00059836
CAS-Nummer: 6461-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR