2,3,4,5-Tetrachloranilin

Lieferant: CHEMPUR

BTB15037.250 BTB15037.1
CHMPBTB15037.250EA 0 EUR
CHMPBTB15037.250 MAYBBTB15037.1
2,3,4,5-Tetrachloranilin
2,3,4,5-Tetrachloranilin
Formel: C₆H₃Cl₄N
Molecular Weight: 230,91 g/mol
CAS-Nummer: 634-83-3
EINECS: 211-216-1
UN: 1648
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR