Dicyclopropylketon

Lieferant: CHEMPUR

Synonyme: Dicyclopropylketone, Dicyclopropylmethanone

DSHS00118.250 DSHS00118.1
CHMPDSHS00118.250EA 0 EUR
CHMPDSHS00118.250 MAYBDSHS00118.1
Dicyclopropylketon
Dicyclopropylketon
Formel: C₇H₁₀O
Molecular Weight: 110,16 g/mol
MDL: MFCD00001276
CAS-Nummer: 1121-37-5
UN: 1224
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR