Methyl-3-isocyanatobenzoat

Lieferant: CHEMPUR

Synonyme: Methyl3-isocyanatobenzoate, 3-(Methoxycarbonyl)phenyl isocyanate

FL13468.POR
CHMPFL13468.POREA 0 EUR
CHMPFL13468.POR
Methyl-3-isocyanatobenzoat
Methyl-3-isocyanatobenzoat
Formel: C₉H₇NO₃
Molecular Weight: 177,16 g/mol
MDL: MFCD00037077
CAS-Nummer: 41221-47-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR