3-Chlorphthalsäureanhydrid

Lieferant: CHEMPUR

Synonyme: 4-Chloroisobenzofuran-1,3-dione

FL13484.POR 013484-5G 013484-25G
CHMPFL13484.POREA 0 EUR
CHMPFL13484.POR FLUO013484-5G FLUO013484-25G
3-Chlorphthalsäureanhydrid
3-Chlorphthalsäureanhydrid
Formel: C₈H₃ClO₃
Molecular Weight: 182,56 g/mol
MDL: MFCD00023107
CAS-Nummer: 117-21-5
EINECS: 204-179-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR