2-Chlor-5-(trifluormethylphenyl)isocyanat
Lieferant: CHEMPUR
Synonyme:
1-Chloro-2-isocyanato-4-(trifluoromethyl)benzene
FL13491.POR
CHMPFL13491.POREA
0
EUR
CHMPFL13491.POR
2-Chlor-5-(trifluormethylphenyl)isocyanat
2-Chlor-5-(trifluormethylphenyl)isocyanat
Formel:
C₈H₃ClF₃NO Molecular Weight: 221,57 g/mol Siedepunkt: 63…63,5 °C (5 mmHg) Dichte: 1,4802 g/cm³ (20 °C) |
MDL:
MFCD00037029 CAS-Nummer: 50528-86-4 UN: 2206 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...