Bromcyclobutan
Lieferant: CHEMPUR
|
Gefahr
|
Synonyme:
Cyclobutylbromid
FL13495.POR
CHMPFL13495.POREA
0
EUR
CHMPFL13495.POR
Bromcyclobutan
Bromcyclobutan
Formel:
C₄H₇Br Molecular Weight: 135 g/mol Siedepunkt: 109…113 °C (1013 hPa) Dichte: 1,4208 g/cm³ (20 °C) |
MDL:
MFCD00001317 CAS-Nummer: 4399-47-7 EINECS: 224-530-9 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...