1,2-Epithiocyclohexan

Lieferant: CHEMPUR

Synonyme: Epithiocyclohexan, Cyclohexensulfid

FL13496.POR
CHMPFL13496.POREA 0 EUR
CHMPFL13496.POR
1,2-Epithiocyclohexan
1,2-Epithiocyclohexane
Formel: C₆H₁₀S
Molecular Weight: 114,21 g/mol
Siedepunkt: 55 °C (1013 hPa)
Dichte: 1,041 g/cm³ (20 °C)
MDL: MFCD00005163
CAS-Nummer: 286-28-2
EINECS: 206-008-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR