(3-Fluor-4-methoxyphenyl)essigsäure

Lieferant: CHEMPUR

Synonyme: 3-Fluoro-4-methoxyphenylacetic acid

FL13841.POR
CHMPFL13841.POREA 0 EUR
CHMPFL13841.POR
(3-Fluor-4-methoxyphenyl)essigsäure
(3-Fluor-4-methoxyphenyl)essigsäure
Formel: C₉H₉FO₃
Molecular Weight: 184,17 g/mol
MDL: MFCD00060353
CAS-Nummer: 452-14-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR