1-(2,5-Dimethoxyphenyl)piperazin

Lieferant: CHEMPUR

FL19969.POR 019969-5G
CHMPFL19969.POREA 0 EUR
CHMPFL19969.POR FLUO019969-5G
1-(2,5-Dimethoxyphenyl)piperazin
1-(2,5-Dimethoxyphenyl)piperazin
Formel: C₁₂H₁₈N₂O₂
MDL: MFCD03444397
CAS-Nummer: 1019-06-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR