Temperature compensator (ATC)
Lieferant: CONSORT
ST10N
CONSST10NEA 135 EUR
CONSST10N
Temperature compensator (ATC)
Temperaturfühler
General purpose temperatur sensor for field use.
  • Stainless steel body, PTFE coated
  • Not suitable for very aggressive fluids