Single use microfibre products, MicroGenesis™
Lieferant: CONTEC
MGMP0001
CONTMGMP0001EA 364 EUR
CONTMGMP0001
Single use microfibre products, MicroGenesis™
Wischtücher Reinigungskits und -Pads
These single use microfibre pads feature synthetic construction for the effective removal of both gross contamination and fine particles. Conveniently designed to provide a new pad for each new use, they are compatible with most existing hardware.
  • Compatible with common disinfectants like quats, peroxides, and bleach
  • Simplifies quality control, inventory, and space management, all without cross contamination risk
  • Provides efficient and effective removal of surface contamination
  • MicroGenesis™ floor pads and MicroGenesis™ hygienic backer plate can be used together
MicroGenesis™ Microfibre Floor Pads
These 100% synthetic, disposable microfibre floor pads effectively remove fine particles and large debris from floors, providing superior removal of visible contaminants. Designed to be used wet or dry for general facility cleaning in ISO Class 8 environments and above. Compatible with a wide range of disinfectants, these synthetic pads are designed to provide uniform surface coverage. A new pad for each application minimises cross-contamination risks (both residual and microbial) unlike laundered mops, and does not interfere with disinfectant efficacy like cotton or rayon. Contec’s hardware can be used for improved functionality and ergonomic ease.