Transport Boxes, Fiberboard
Lieferant: E3 Cortex
3CAH000981
CORT3CAH000981EA 63.2 EUR
CORT3CAH000981
Transport Boxes, Fiberboard
Boxen Transportboxen
These cardboards boxes are used for the transfer of dangerous and\or regulated goods by air, road and maritime traffic. They allow the shipping of liquid products or solids of any classes (except 6.2 and 7) according to regulations in force (IATA, ADR, IMDG, RID).
  • Assembly in two minutes
  • Absorbent vermiculite for dangerous products of the groups of Packaging I, II and III
  • All the references are UN certified 4GV, allowing the use of every type of internal bowl
  • Must be used with an absorbent
Standards: UN certified.
Verpackung: Delivered with plastic bag.