Concrete D-ring anchor, DBI-SALA, 3M™
Lieferant: CAPITAL SAFETY
2104565
CPSD2104565EA 0 EUR
CPSD2104565
Concrete D-ring anchor, DBI-SALA, 3M™
Fallschutz Höhensicherungsgeräte und Fallbegrenzer
Reusable D-Ring assembly in a rugged design for harsh environments.
  • Safe 'hook-up' indicator notifies user of proper installation
  • For 18 or 19 mm holes - only standard drill bits needed
Standards: EN 795, Class B