Fixed anchorage plate, Protecta, 3M™
Lieferant: CAPITAL SAFETY
AM210
CPSDAM210EA 0 EUR
CPSDAM210
Fixed anchorage plate, Protecta, 3M™
Fallschutz Höhensicherungsgeräte und Fallbegrenzer
Protecta® Fixed anchorage plate made from stainless steel.
  • 12 mm fixing
  • 3 mm thick
Standards: EN 795, Class A
Lieferumfang: Supplied without fixing.