Supelco® Organic Reference Standard, F.A.M.E. Mix GLC-10
Lieferant: Merck
CRM1891
CRM1891EA 61.9 EUR
CRM1891
Supelco® Organic Reference Standard, F.A.M.E. Mix GLC-10
Standards Organische Standards
This certified reference material is useful for determining relative retention times and approximating response factors.