Direct Blue 15, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
D2535-25G
D2535-25GEA
148
EUR
D2535-25G
Direct Blue 15, Sigma-Aldrich®
Direct Blue 15
May be used as a substitute for trypan blue for some biological and staining applications.
Formel:
C₃₄H₂₄N₆Na₄O₁₆S₄ Molecular Weight: 992,8 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036441 CAS-Nummer: 2429-74-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...