o-Dianisidin Dihydrochlorid, Supelco®
Lieferant: Merck
Synonyme:
3,3'-Dimethoxybenzidine dihydrochloride
M-Clarity™ Qualitätsstufe = MQ100
D2679-1VL
D2679-1VLEA
52.7
EUR
D2679-1VL
o-Dianisidin Dihydrochlorid, Supelco®
o-Dianisidin Dihydrochlorid
Formel:
C₁₄H₁₈Cl₂N₂O₂ Molecular Weight: 317,21 g/mol |
MDL:
MFCD00012488 CAS-Nummer: 20325-40-0 EINECS: 243-737-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...