Safety Bag, Bubble
Lieferant: DAKLAPACK
460698
DAKL460698EA 156 EUR
DAKL460698
Safety Bag, Bubble
Beutel Aufbewahrungsbeutel
Semi transparent PE bubble foil safety bag with closure type-destructive tape.
  • Can be used in conjunction with the Minimailbox, absorbing material and Transportblister
Fitted with the easily identifiable blue band across the top of the bag which when folded back exposes the peel away backing, simply insert contents into the Safetybag, removing backing and apply even pressure across the blue band surface. The Safetybag will now be completely leak proof. Suitable as a secondary liquid tight packaging for transport of biological substances category B.
Standards: Certified with P650, UN3373.