Multiple use earplugs, Quiet®
Lieferant: HONEYWELL SAFETY
1028456
DALL1028456EA 63.6 EUR
DALL1028456
Multiple use earplugs, Quiet®
Gehörschutzstöpsel Gehörschutzstöpsel, wiederverwendbar
These vinyl multiple earplugs are available with or without cord.
  • Bell shape: 'no-roll' design that is easy to handle and fit
  • Contoured shape comfortably matches contours of the ear canal
  • Smooth, non-irritating skin provides all-day comfort, easy to clean for long-term use
  • Built-in insertion stem makes insertion quick and easy
Suitable for use in chemical, manufacturing, medical, mining, municipal services, pharmaceutical and transportation fields.
SNR: 28 dB
NRR: 26
Standards: EN 352-2