N-Octyl-β-D-glucopyranosid
Lieferant: G Bioscienes
|
Achtung
|
Synonyme:
Octyl-β-D-glucosid, Octyl-β-D-glucopyranosid
DG016
DG016EA
1290
EUR
DG016
N-Octyl-β-D-glucopyranosid
N-Octyl-β-D-glucopyranosid
Formel:
C₁₄H₂₈O₆ Molecular Weight: 292,37 g/mol Schmelzpunkt: 110 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00063288 CAS-Nummer: 29836-26-8 EINECS: 249-887-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...