Basic spectrometer, mini-SPID
Lieferant: DIDALAB
POF010350
DIDAPOF010350EA 1400 EUR
DIDAPOF010350
Basic spectrometer, mini-SPID
Spektrophotometer und Photometer UV/Vis-Spektralphotometer
mini-SPID is a pedagogical spectrometer used in the visible range.
To analyse a signal in real time of transmission, ray and continuous spectra (sun, glowing lamp, candle), detections and photometric measures in high schools.
Lieferumfang: Supplied with intuitive software, two meter optical fibre cable and USB cord.