Dewar vessel calorimeter
Lieferant: DIDALAB
PTD039501
DIDAPTD039501EA 583 EUR
DIDAPTD039501
Dewar vessel calorimeter
Kalorimeter
This calorimeter includes an aluminium container of 500 ml, concentric to one large neck Dewar vase. The metal internal vase is supported by plastic ring, laying on the Dewar vessel large neck. The set is protected by a metal casing, equipped with two locking hooks.
White internal plastic cover including a square section opening for the stirring rod, blocked in rotation because of the square section of the rod.
Central closing with Ø 40 mm protection cap, for isolation of foreign bodies after having been introduced into the calorimeter and a thermometer of Ø 8 mm opening.
Cover with rubber washer hold by two metal clamp collars diametrically opposed (outer Ø of 170 mm, height of 215 mm).
Lieferumfang: Supplied with glass beaker 500 ml and handbook.