Thermal transfer Cryo-Tags® labels
Lieferant: DIVERSIFIED BIOTECH
CRTH-2000
DIVECRTH-2000EA 557 EUR
DIVECRTH-2000
Thermal transfer Cryo-Tags® labels
Etiketten
These Cryo-Tags® polyester labels work with most thermal transfer printers to print popular barcode formats for fast and easy sample identification.
  • Temperature resistant from −196 to +80 °C
  • Freezable in liquid and vapour phase nitrogen
  • Solvent-resistant, acrylic-based adhesive provides excellent cold temperature performance
  • Resists degradation from scuffing, chemicals, moisture and temperature fluctuations
Print crisp, clear, professional quality labels.