Counting chamber with grid
Lieferant: DIVERSIFIED BIOTECH
GRID-1000
DIVEGRID-1000EA 138 EUR
DIVEGRID-1000
Counting chamber with grid
Objektträger Zählkammern
Disposable polyester with acrylic adhesive grid stickers for micro or semi-micro objects.
  • Inert and stable under most conditions
  • Compatible with different kinds of microscopes
  • Separate coverslips can be placed over each sticker
These slide grids are either used with dry or suspended specimens, such as cells held in buffer, under cover slips.