Waschmittel, konzentriert, Shureclean Plus VK9
Lieferant: Diversey
7509854
DIVY7509854EA 75.9 EUR
DIVY7509854
Waschmittel, konzentriert, Shureclean Plus VK9
Reinigungsmittel Waschmittel und Oberflächenreiniger
Shureclean Plus VK9 is a concentrated, neutral, high foaming liquid detergent designed for general purpose applications in the food, beverage and dairy industries. This detergent is recommended for manual and soak cleaning of all food preparation equipment, cooking vessels and utensils. It can also be used for cleaning floors, walls and work surfaces.
  • Clear, pale brown liquid
  • Relative density at 20 °C - 1,03
  • Suitable for soft or hard water and pH neutral
  • Non perfumed product