Safety and Cleaning Container
Lieferant: DUPERTHAL
3.89.005
DUEH3.89.005EA 226.28 EUR
DUEH3.89.005
Safety and Cleaning Container
Transportbehälter Flaschentragebehälter
Unpolished, stainless steel 1.4571 Dimensiom: 165×380 (Ø×H) mm tigth closure, Captive cap on flexible shaft Flame arrester in can neck.
Standards: Type approval tested can according to ADR, RID and IMDG code Markings prepared according to GefStoffV and BGV A8 Aditional markings according to ADR/RID Testing: BAM-admittance.