Organic reference standard, Endotoxin standard BRP, Ph.Eur., RS
Lieferant: EDQM
E0150000
EDQME0150000EA 245 EUR
EDQME0150000
Organic reference standard, Endotoxin standard BRP, Ph.Eur., RS
Standards Organische Standards
  • In Monographs 20614 and 20630
  • Freeze-dried material