Organic reference standard, Ethinylestradiol, Ph.Eur., RS
Lieferant: EDQM
E1900000
EDQME1900000EA 245 EUR
EDQME1900000
Organic reference standard, Ethinylestradiol, Ph.Eur., RS
Standards Organische Standards
Filled under inert atmosphere protected from humidity.
  • In Monograph 0140, 0821, 0940, and 0926