Pharmaceutical Analytical Standard, Escitalopram for System Suitability, Reference Material
Lieferant: EDQM
Y0001797
EDQMY0001797EA 197 EUR
EDQMY0001797
Pharmaceutical Analytical Standard, Escitalopram for System Suitability, Reference Material
Standards Organische Standards
Escitalopram for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.
Escitalopram is a selective serotonin reuptake inhibitor (SSRI), the S-enantiomer and eutomer of citalopram.