X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)

Lieferant: EDVOTEK

Synonyme: BCIG

614
EDVO614EA 44 EUR
EDVO614
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
Formel: C₁₄H₁₅BrClNO₆
Molecular Weight: 408,63 g/mol
Schmelzpunkt: 237…239 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00005666
CAS-Nummer: 7240-90-6
EINECS: 230-640-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR