1,3-Diphenylharnstoff

Lieferant: EHRENSTORFER

Synonyme: N,N’-Diphenylurea, N,N'-Diphenylurea

C12921500
EHERC12921500EA 44 EUR
EHERC12921500
1,3-Diphenylharnstoff
1,3-Diphenylharnstoff
Formel: C₁₃H₁₂N₂O
Molecular Weight: 212,25 g/mol
MDL: MFCD00003017
CAS-Nummer: 102-07-8
EINECS: 203-003-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR