Flurprimidol
Lieferant: EHRENSTORFER
C13851000
EHERC13851000EA
150
EUR
EHERC13851000
Flurprimidol
Flurprimidol
Formel:
C₁₅H₁₅F₃N₂O₂ |
CAS-Nummer:
56425-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...