17α-Hydroxyprogesteron
Lieferant: EHRENSTORFER
Synonyme:
17α-Hydroxy-4-pregnene-3,20-dione
C14241000
EHERC14241000EA
45
EUR
EHERC14241000
17α-Hydroxyprogesteron
17α-Hydroxyprogesteron
Formel:
C₂₁H₃₀O₃ Molecular Weight: 330,46 g/mol |
MDL:
MFCD00003659 CAS-Nummer: 68-96-2 EINECS: 200-699-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...