Sulfachinoxalin
Lieferant: EHRENSTORFER
Synonyme:
4-Amino-N-(quinoxalin-2-yl)benzenesulphonamide
C16990000
EHERC16990000EA
60
EUR
EHERC16990000
Sulfachinoxalin
Sulfachinoxalin
Formel:
C₁₄H₁₂N₄O₂S Molecular Weight: 300,3357 g/mol |
MDL:
MFCD00055406 CAS-Nummer: 59-40-5 EINECS: 200-423-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...