Disinfectant tablets
Lieferant: ELGA LabWater
CT3 CT3-50
ELGACT3EA 55 EUR
ELGACT3 ELGACT3-50
Disinfectant tablets
Reinigungsmittel Desinfektionsmittel
These disinfectant tablets sanitises tubing or pipe work to remove bacteria, reduce bio film and protect the system against bacterial growth.