Drums, HDPE, FluoroPure®
Lieferant: ENTEGRIS
4B0100-0006
ENTE4B0100-0006EA 0 EUR
ENTE4B0100-0006
Drums, HDPE, FluoroPure®
Fässer, Fässer, Trommeln
HDPE, triple layer.
  • Single-piece design with molded-in chime increases strength and closure protection
  • UN tested and approved
  • Pressure tested and assembled in Class 6 cleanroom ensuring a high purity package
  • Accommodates standard Sentry® drum insert length
  • Not recommended for pressure dispense or vacuum conditions
Designed with multilayer which includes a wetted inner layer of virgin HDPE and is ideal for maximum retrevability with a unique sump that minimizes residual chemical. The port sealing surface allows FEP encapsulated silicone O rings to provide closure seal.