In-line gauge protector
Lieferant: ENTEGRIS
IGP-02-060-4-6
ENTEIGP-02-060-4-6EA 0 EUR
ENTEIGP-02-060-4-6
In-line gauge protector
Pumpen Vakuum-Messgeräte
Isolates corrosive chemicals from gauges.
  • All wetted surfaces are of molded PFA material
  • Exterior gauge protector body surfaces are PFA and PVDF material
  • Full flow-through design
  • Gauges consist of a 2 ¹/₂" (63,5 mm) dial, with stainless steel case and stainless steel movement
  • Accepts most ¹/₄" MNPT gauge and transducer designs