Savana® PTFE Disposable Filters
Lieferant: ENTEGRIS
SVEADS2N1
ENTESVEADS2N1EA 0 EUR
ENTESVEADS2N1
Savana® PTFE Disposable Filters
Filter Membranfilter
Savana® PTFE disposable filters are constructed of PTFE pleated membranes with polypropylene supports and housing to provide fine durability and cleanliness for filtration of process fluids used in the electronics and various industries.
  • The hydrophobic version can be used in IPA or weak solvent applications in flat panel display or hard disk industries
  • Pleated PTFE membrane with polypropylene support and housing
  • Better particle removal performance than polypropylene media
  • Excellent chemical compatibility
  • Suitable for various formats and flow requirements