Glutathione peroxidase activity kit
Lieferant: ENZO LIFE SCIENCES AG
ADI900158
ENZOADI900158EA 848.1 EUR
ENZOADI900158
Glutathione peroxidase activity kit
Assays Enzymtests
The Glutathione Peroxidase activity kit is a convenient 96-well plate based assay for the higher throughput analysis of glutathione peroxidase activity in a variety of sample matrices in just 30 minutes.
For the measurement of glutathione peroxidase in cell lysates, plasma, and tissue from any species.
Bestellinformation: Contents: Microtiter plate, Assay buffer concentrate, Reaction mix, Glutathione reductase, GSSG