Glutathione peroxidase activity kit
Lieferant: ENZO LIFE SCIENCES AG
ADI900158
ENZOADI900158EA
848.1
EUR
ENZOADI900158
Glutathione peroxidase activity kit
Assays
Enzymtests
The Glutathione Peroxidase activity kit is a convenient 96-well plate based assay for the higher throughput analysis of glutathione peroxidase activity in a variety of sample matrices in just 30 minutes.
For the measurement of glutathione peroxidase in cell lysates, plasma, and tissue from any species.
Bestellinformation: Contents: Microtiter plate, Assay buffer concentrate, Reaction mix, Glutathione reductase, GSSG