Ampicillin Natriumsalz ≥93%

Lieferant: ENZO LIFE SCIENCES AG

Synonyme: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt

ALX380268G005
ENZOALX380268G005EA 72.6 EUR
ENZOALX380268G005
Ampicillin Natriumsalz ≥93%
Ampicillin sodium salt

β-Lactam antibiotic. Inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking). Inactivates transpeptidases on the inner surface of the bacterial cell membrane. Effective against Gram-negative and Gram-positive bacteria.


  • Semisynthetic from penicillin
  • Inactivated by β-lactamase


White to light yellow powder


Soluble in water (50 mg/mL)


Verpackung: Shipping temperature: Ambient

Formel: C₁₆H₁₈N₃NaO₄S
Molecular Weight: 371,39 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00064313
CAS-Nummer: 69-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR